missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ MK 1903

Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Description
Potent and selective hydroxycarboxylic acid receptor 2 (HCA2, GPR109A) full agonist; exhibits greater potency than niacin in a whole cell HTRF-cAMP assay (EC50 values are 12.9 and 51 nM respectively). Exhibits no binding at the GRP109B receptor.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 164.16 |
| Physical Form | Solid |
| Color | White |
| Purity | >98% |
| Molecular Weight (g/mol) | 164.164 |
| Chemical Name or Material | MK 1903 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction