missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLH3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-04481-100UL
This item is not returnable.
View return policy
Description
MLH3 Polyclonal specifically detects MLH3 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.
Specifications
| MLH3 | |
| Polyclonal | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| DNA mismatch repair protein Mlh3, HNPCC7, MGC138372, mutL (E. coli) homolog 3, mutL homolog 3 (E. coli), MutL protein homolog 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1180-1429 of human MLH3 (NP_055196.2). | |
| 100 μL | |
| DNA Repair, Mismatch Repair | |
| 27030 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction