missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MLH3 Rabbit anti-Human, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 190.00 - € 482.00
Specifications
| Antigen | MLH3 |
|---|---|
| Applications | Western Blot, Immunofluorescence |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18341916
|
Novus Biologicals
NBP3-04481-20UL |
20 μL |
€ 190.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18083554
|
Novus Biologicals
NBP3-04481-100UL |
100 μL |
€ 482.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MLH3 Polyclonal specifically detects MLH3 in Human samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
| MLH3 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| DNA mismatch repair protein Mlh3, HNPCC7, MGC138372, mutL (E. coli) homolog 3, mutL homolog 3 (E. coli), MutL protein homolog 3 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1180-1429 of human MLH3 (NP_055196.2). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| Rabbit | |
| DNA Repair, Mismatch Repair | |
| PBS with 50% glycerol, pH7.3. | |
| 27030 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title