missing translation for 'onlineSavingsMsg'
Learn More
Learn More
COMT Rabbit anti-Human, Rat, Clone: 9Y2A3, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Bio-Techne NBP3-16413-100UL
This item is not returnable.
View return policy
Description
COMT Monoclonal antibody specifically detects COMT in Human, Rat samples. It is validated for Western Blot
Specifications
| COMT | |
| Monoclonal | |
| Unconjugated | |
| catechol O-methyltransferase, catechol-O-methyltransferase, EC 2.1.1.6 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of human COMT (P21964). | |
| 100 μg | |
| Lipid and Metabolism | |
| 1312 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| 9Y2A3 | |
| PBS, 0.05% BSA, 50% glycerol, pH7.3 | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction