missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-1 Antibody (SR1951), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22010-100ul
This item is not returnable.
View return policy
Description
MMP-1 Monoclonal antibody specifically detects MMP-1 in Human samples. It is validated for Western Blot, Immunoprecipitation
Specifications
| MMP-1 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot, Immunoprecipitation | |
| SR1951 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| CLGmatrix metalloprotease 1, CLGN, EC 3.4.24, EC 3.4.24.7, Fibroblast collagenase, interstitial collagenase, matrix metallopeptidase 1 (interstitial collagenase), matrix metalloproteinase 1 (interstitial collagenase), Matrix metalloproteinase-1, MMP-1 | |
| A synthesized peptide derived from human MMP-1 (Uniprot #: P03956) | |
| 100 μg | |
| Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix | |
| 4312 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction