missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-1 Antibody (SR1951), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 338.00 - € 449.00
Specifications
| Antigen | MMP-1 |
|---|---|
| Clone | SR1951 |
| Dilution | Western Blot 1:500-1:2000, Immunoprecipitation 1:50 |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18669184
|
Novus Biologicals
NBP3-22010-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18677504
|
Novus Biologicals
NBP3-22010-50ul |
50 μg |
€ 338.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MMP-1 Monoclonal antibody specifically detects MMP-1 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
| MMP-1 | |
| Western Blot 1:500-1:2000, Immunoprecipitation 1:50 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CLGmatrix metalloprotease 1, CLGN, EC 3.4.24, EC 3.4.24.7, Fibroblast collagenase, interstitial collagenase, matrix metallopeptidase 1 (interstitial collagenase), matrix metalloproteinase 1 (interstitial collagenase), Matrix metalloproteinase-1, MMP-1 | |
| A synthesized peptide derived from human MMP-1 (Uniprot #: P03956) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1951 | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| Rabbit | |
| Angiogenesis, Cancer, Cell Biology, Cellular Markers, Extracellular Matrix | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 4312 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title