missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-7 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-99123SS
This item is not returnable.
View return policy
Description
MMP-7 Polyclonal specifically detects MMP-7 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| MMP-7 | |
| Polyclonal | |
| Unconjugated | |
| PBS (pH 7.4) with 0.02% Sodium Azide | |
| EC 3.4.24, EC 3.4.24.23, matrilysin, matrin, matrix metallopeptidase 7 (matrilysin, uterine), matrix metalloproteinase 7 (matrilysin, uterine), Matrix metalloproteinase-7, MMP-7, MPSL1, PUMP1, PUMP-1, Pump-1 protease, uterine matrilysin, Uterine metalloproteinase | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| 1.28 mg/mL | |
| Immunohistochemistry 1:300, Immunohistochemistry-Paraffin 1:300 | |
| P09237 | |
| MMP7 | |
| A synthetic peptide made to an internal portion of the human MMP7 protein (between residues 250-300) [UniProt P09237] | |
| 0.025 mL | |
| Angiogenesis, Cancer, Cell Biology, Cytoskeleton Markers, Extracellular Matrix, Wnt Signaling Pathway | |
| 4316 | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction