missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MMP-7 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 208.00 - € 473.00
Specifications
| Antigen | MMP-7 |
|---|---|
| Concentration | 1.28 mg/mL |
| Dilution | Immunohistochemistry 1:300, Immunohistochemistry-Paraffin 1:300 |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18043857
|
Novus Biologicals
NBP1-99123SS |
0.025 mL |
€ 208.00
25µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18052567
|
Novus Biologicals
NBP1-99123 |
0.1 mL |
€ 473.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MMP-7 Polyclonal specifically detects MMP-7 in Human, Mouse samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| MMP-7 | |
| Immunohistochemistry 1:300, Immunohistochemistry-Paraffin 1:300 | |
| Polyclonal | |
| Rabbit | |
| Angiogenesis, Cancer, Cell Biology, Cytoskeleton Markers, Extracellular Matrix, Wnt Signaling Pathway | |
| PBS (pH 7.4) with 0.02% Sodium Azide | |
| EC 3.4.24, EC 3.4.24.23, matrilysin, matrin, matrix metallopeptidase 7 (matrilysin, uterine), matrix metalloproteinase 7 (matrilysin, uterine), Matrix metalloproteinase-7, MMP-7, MPSL1, PUMP1, PUMP-1, Pump-1 protease, uterine matrilysin, Uterine metalloproteinase | |
| MMP7 | |
| IgG | |
| Affinity Purified |
| 1.28 mg/mL | |
| Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| Human, Mouse | |
| P09237 | |
| 4316 | |
| A synthetic peptide made to an internal portion of the human MMP7 protein (between residues 250-300) [UniProt P09237] | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title