missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Bovine IgG1 Heavy Chain Secondary Antibody (IL-A60), Janelia Fluor 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-68415JF646
This item is not returnable.
View return policy
Description
IgG1 Heavy Chain Monoclonal antibody specifically detects IgG1 Heavy Chain in Bovine samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen)
Specifications
| IgG1 Heavy Chain | |
| Monoclonal | |
| Janelia Fluor 646 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Bovine | |
| Purified |
| Flow Cytometry, ELISA, Immunohistochemistry, Immunoprecipitation, Immunohistochemistry (Frozen) | |
| IL-A60 | |
| 50mM Sodium Borate | |
| Purified bovine immunoglobulin | |
| 0.1 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction