missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Bovine IgG2 Heavy Chain Secondary Antibody (IL-A73), PerCP, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-68417PCP
This item is not returnable.
View return policy
Description
IgG2 Heavy Chain Monoclonal antibody specifically detects IgG2 Heavy Chain in Bovine samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation
Specifications
| IgG2 Heavy Chain | |
| Monoclonal | |
| PerCP | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Bovine | |
| Purified |
| Flow Cytometry, ELISA, Immunoprecipitation | |
| IL-A73 | |
| PBS | |
| Purified bovine IgG2 | |
| 0.1 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction