missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Bovine IgM (H+L) Secondary Antibody (IL-A30), FITC, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-68419F
This item is not returnable.
View return policy
Description
IgM (H+L) Monoclonal antibody specifically detects IgM (H+L) in Bovine samples. It is validated for Flow Cytometry, ELISA, Immunoprecipitation, Radioimmune Assays
Specifications
| IgM (H+L) | |
| Monoclonal | |
| FITC | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Bovine | |
| Purified |
| Flow Cytometry, ELISA, Immunoprecipitation, Radioimmune Assays (RIA) | |
| IL-A30 | |
| PBS | |
| Bovine peripheral blood mononuclear cells depleted of T lymphocytes | |
| 0.1 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction