missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Canine IgA/IgG/IgM (H+L) Secondary Antibody (K99 2E3), DyLight 594, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP3-11626DL594
This item is not returnable.
View return policy
Description
IgA/IgG/IgM (H+L) Monoclonal antibody specifically detects IgA/IgG/IgM (H+L) in Canine samples. It is validated for ELISA
Specifications
| IgA/IgG/IgM (H+L) | |
| Monoclonal | |
| DyLight 594 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Canine | |
| Purified |
| ELISA | |
| K99 2E3 | |
| 50mM Sodium Borate | |
| Canine immunoglobulin | |
| 0.1 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction