missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgE Heavy Chain Secondary Antibody (E454 (5D5)), DyLight 680, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-68297FR
This item is not returnable.
View return policy
Description
IgE Heavy Chain Monoclonal antibody specifically detects IgE Heavy Chain in Human samples. It is validated for ELISA
Specifications
| IgE Heavy Chain | |
| Monoclonal | |
| DyLight 680 | |
| Constant Region of Heavy Chain of IgE, Ig Epsilon Chain C Region, Ig Epsilon Chain C Region ND, IgE, Immunoglobulin Epsilon, immunoglobulin heavy constant epsilon | |
| Immunoglobulin E from human serum | |
| 0.2 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG2a |
| ELISA | |
| E454 (5D5) | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein G purified | |
| RUO | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction