Learn More
Mouse anti-Human IgG Fc Secondary Antibody (8A4), Janelia Fluor 549, Novus Biologicals™
Mouse Monoclonal Antibody
Brand:  Novus Biologicals NBP2-68421JF549
Product Code. 30232652
  • € 748.00 / 0.20mL
Description
IgG Fc Monoclonal antibody specifically detects IgG Fc in Human samples. It is validated for Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IgG Fc
Monoclonal
Janelia Fluor 549
Mouse
Protein A purified
RUO
Human
Purified
Flow Cytometry, ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
8A4
50mM Sodium Borate
Human IgG polyclonal
0.2 mL
Secondary
Store at 4°C in the dark.