missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG Fc Secondary Antibody (EM-07), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-45063R
This item is not returnable.
View return policy
Description
IgG Fc Monoclonal antibody specifically detects IgG Fc in Human samples. It is validated for Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence
Specifications
| IgG Fc | |
| Monoclonal | |
| DyLight 550 | |
| Mouse | |
| Protein A purified | |
| RUO | |
| Human | |
| Purified |
| Western Blot, Flow Cytometry, ELISA, Immunocytochemistry/Immunofluorescence | |
| EM-07 | |
| 50mM Sodium Borate | |
| Fusion protein of human IgG Fc fragment. | |
| 0.1 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction