missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Human IgG Secondary Antibody (MR12) (BSA Free), Novus Biologicals™
Mouse Monoclonal Antibody
€ 215.00 - € 392.00
Specifications
| Antigen | IgG |
|---|---|
| Clone | MR12 |
| Concentration | 1 mg/mL |
| Dilution | Immunohistochemistry 1:10 - 1:500 |
| Applications | Immunohistochemistry |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18607927
|
Novus Biologicals
NBP2-50261-0.025ml |
0.025 mL |
€ 215.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18667526
|
Novus Biologicals
NBP2-50261 |
0.1 mL |
€ 392.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
IgG Monoclonal antibody specifically detects IgG in Human samples. It is validated for ImmunohistochemistrySpecifications
| IgG | |
| 1 mg/mL | |
| Immunohistochemistry | |
| Unconjugated | |
| Mouse | |
| Human | |
| Immunoglobulin G, ImmunoglobulinG | |
| Immunoglobulin purified from rabbit serum | |
| Secondary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| MR12 | |
| Immunohistochemistry 1:10 - 1:500 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS | |
| 3500 | |
| IgG1 | |
| Protein A purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title