missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Rabbit IgG (H+L) Secondary Antibody, DyLight 755 (Pre-adsorbed), Novus Biologicals™
Mouse Polyclonal Antibody
Brand: Novus Biologicals NBP1-72764IR
This item is not returnable.
View return policy
Description
IgG (H+L) Polyclonal antibody specifically detects IgG (H+L) in Rabbit samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Dot Blot
Specifications
| IgG (H+L) | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Mouse anti-Rabbit IgG (H+L) Secondary Antibody (Pre-adsorbed) was produced by repeated immunization with rabbit whole IgG molecule in mouse. | |
| 0.5 mL | |
| Adaptive Immunity, Immunology | |
| Rabbit | |
| IgG |
| Western Blot, ELISA, Immunohistochemistry, Dot Blot | |
| DyLight 755 | |
| Mouse | |
| Multi-step | |
| RUO | |
| Secondary | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction