missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Rat IgA Heavy Chain Secondary Antibody (MARA-1), DyLight 488, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-68392G
This item is not returnable.
View return policy
Description
IgA Heavy Chain Monoclonal antibody specifically detects IgA Heavy Chain in Rat samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunofluorescence
Specifications
| IgA Heavy Chain | |
| Monoclonal | |
| DyLight 488 | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Rat | |
| Purified |
| ELISA, Immunohistochemistry, Immunohistochemistry (Frozen), Immunofluorescence | |
| MARA-1 | |
| 50mM Sodium Borate | |
| Purified IR1060 IgA rat myeloma protein | |
| 0.25 mL | |
| Secondary | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction