missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Rat IgD Secondary Antibody (MARD-3), DyLight 405, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-68388V
This item is not returnable.
View return policy
Description
IgD Monoclonal antibody specifically detects IgD in Rat samples. It is validated for ELISA, Immunohistochemistry, Immunohistochemistry (Frozen)
Specifications
| IgD | |
| Monoclonal | |
| DyLight 405 | |
| IgD, Igh-5, Ighd, Immunoglobulin D, Immunoglobulin Delta | |
| IR731 myeloma protein | |
| 0.25 mL | |
| B Cell Development and Differentiation Markers, Immunology | |
| 3495 | |
| Store at 4°C in the dark. | |
| IgG1 |
| ELISA, Immunohistochemistry, Immunohistochemistry (Frozen) | |
| MARD-3 | |
| 50mM Sodium Borate | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction