missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Rat IgE Heavy Chain Secondary Antibody (MARE-1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-68394-250UG
This item is not returnable.
View return policy
Description
IgE Heavy Chain Monoclonal specifically detects IgE Heavy Chain in Rat samples. It is validated for Western Blot, ELISA.
Specifications
| IgE | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot, ELISA | |
| IgE, immunoglobulin heavy constant epsilon | |
| Mouse | |
| Affinity Purified | |
| Adaptive Immunity, Immunology | |
| Mouse anti Rat IgE antibody, clone MARE-1, recognizes rat epsilon heavy chain of immunoglobulin IgE and does not cross-react with other classes of rat immunoglobulin. The antibody binds to rat IgE with an avidity of 4x10^9 M^-1 | |
| Store at 4°C. |
| Western Blot, ELISA | |
| MARE-1 | |
| Unconjugated | |
| PBS | |
| IGHE | |
| Rat IR162, IR1016, IR2 and IR410 IgE myeloma proteins. | |
| 250 μg | |
| 3497 | |
| Rat | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction