Learn More
Invitrogen™ Mouse anti-Rat IgM (Heavy chain) Secondary Antibody, FITC
Mouse Monoclonal Secondary Antibody
Brand:  Invitrogen™ MA516821
Product Code. 15216069
  • € 768.00 / 500µg
Description
For FACS analysis, use 50 μL of the suggested working dilution to label 1x10^6 cells in 100 μL. Mouse anti Rat IgM Heavy Chain antibody, clone MARM-4 recognizes the mu heavy chain of rat immunoglobulin and does not cross-react with other immunoglobulin classes.
Anti-Rat secondary antibodies are affinity-purified antibodies with well-characterized specificity for rat immunoglobulins and are useful in the detection, sorting or purification of its specified target. Secondary antibodies offer increased versatility enabling users to use many detection systems (e.g. HRP, AP, fluorescence). They can also provide greater sensitivity through signal amplification as multiple secondary antibodies can bind to a single primary antibody. Most commonly, secondary antibodies are generated by immunizing the host animal with a pooled population of immunoglobulins from the target species and can be further purified and modified (i.e. immunoaffinity chromatography, antibody fragmentation, label conjugation, etc.) to generate highly specific reagents.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Rat IgM (Heavy chain)
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide
Polyclonal rat IgM.
500 μg
Secondary
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Liquid
Flow Cytometry
MARM-4
FITC
Mouse
Affinity chromatography
RUO
Rat
Secondary Antibody