missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Rat IgM Heavy Chain Secondary Antibody (HIS40), DyLight 550, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-43375R
This item is not returnable.
View return policy
Description
IgM Heavy Chain Monoclonal antibody specifically detects IgM Heavy Chain in Rat samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| IgM Heavy Chain | |
| Monoclonal | |
| DyLight 550 | |
| Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain | |
| Rat IgM | |
| 0.1 mL | |
| B Cell Development and Differentiation Markers, Immunology | |
| 3507 | |
| Store at 4°C in the dark. | |
| IgG1 κ |
| Flow Cytometry, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| HIS40 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Secondary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction