missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse anti-Rat IgM Heavy Chain Secondary Antibody (MARM-4), Janelia Fluor 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-68381JF646
This item is not returnable.
View return policy
Description
IgM Heavy Chain Monoclonal antibody specifically detects IgM Heavy Chain in Rat samples. It is validated for ELISA
Specifications
| IgM Heavy Chain | |
| Monoclonal | |
| Janelia Fluor 646 | |
| Constant region of heavy chain of IgM, Ig Mu Chain C Region BOT, Ig Mu Chain C Region GAL, Ig Mu Chain C Region OU, immunoglobulin heavy constant mu, Immunoglobulin mu chain | |
| IR202 rat IgM myeloma protein | |
| 0.25 mL | |
| B Cell Development and Differentiation Markers, Immunology | |
| 3507 | |
| Store at 4°C in the dark. | |
| IgG1 |
| ELISA | |
| MARM-4 | |
| 50mM Sodium Borate | |
| Mouse | |
| Affinity purified | |
| RUO | |
| Secondary | |
| Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction