missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse PCPE-1 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF2239
This item is not returnable.
View return policy
Description
PCPE-1/PCOLCE Polyclonal specifically detects PCPE-1/PCOLCE in Mouse samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| PCPE-1/PCOLCE | |
| Polyclonal | |
| Unconjugated | |
| Q61398 | |
| PCOLCE | |
| Mouse myeloma cell line NS0-derived recombinant mouse PCPE-1 Gln25-Ala468 (predicted) Accession # Q61398 | |
| 100 μg | |
| Primary | |
| Detects mouse PCPE-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 35% cross-reactivity with recombinant human PCPE is observed. | |
| Mouse | |
| IgG |
| Western Blot, Immunoprecipitation | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| PCOLCE, PCPE, PCPE1, PCPE-1, PCPE1Type I procollagen COOH-terminal proteinase enhancer, procollagen C-endopeptidase enhancer, procollagen C-endopeptidase enhancer 1, Procollagen COOH-terminal proteinase enhancer 1, Procollagen C-proteinase enhancer 1, procollagen, type 1, COOH-terminal proteinase enhancer, Type 1 procollagen C-proteinase enhancer protein | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5118 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction