missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse PCPE-1 Antibody, R&D Systems™

Goat Polyclonal Antibody
€ 209.00 - € 512.00
Specifications
| Antigen | PCPE-1/PCOLCE |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL |
| Applications | Western Blot, Immunoprecipitation |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18787907
![]() |
R&D Systems
AF2239-SP |
25 μg |
€ 209.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18070492
|
R&D Systems
AF2239 |
100 μg |
€ 512.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
PCPE-1/PCOLCE Polyclonal specifically detects PCPE-1/PCOLCE in Mouse samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| PCPE-1/PCOLCE | |
| Western Blot 0.1 ug/mL, Immunoprecipitation 25 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| PCOLCE, PCPE, PCPE1, PCPE-1, PCPE1Type I procollagen COOH-terminal proteinase enhancer, procollagen C-endopeptidase enhancer, procollagen C-endopeptidase enhancer 1, Procollagen COOH-terminal proteinase enhancer 1, Procollagen C-proteinase enhancer 1, procollagen, type 1, COOH-terminal proteinase enhancer, Type 1 procollagen C-proteinase enhancer protein | |
| PCOLCE | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| LYOPH | |
| Western Blot, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| Q61398 | |
| 5118 | |
| Mouse myeloma cell line NS0-derived recombinant mouse PCPE-1 Gln25-Ala468 (predicted) Accession # Q61398 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse PCPE-1 in direct ELISAs and Western blots. In direct ELISAs and Western blots, approximately 35% cross-reactivity with recombinant human PCPE is observed. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title