missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Plasminogen Kringle 5 Antibody, R&D Systems™
Goat Polyclonal Antibody
Brand: R&D Systems AF742
This item is not returnable.
View return policy
Description
Plasminogen Kringle 5 Polyclonal specifically detects Plasminogen Kringle 5 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| Plasminogen Kringle 5 | |
| Polyclonal | |
| Unconjugated | |
| AAA50168 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 5340 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Plg | |
| E. coli-derived recombinant mouse Plasminogen Kringle 5 Cys481-Ala563 Accession # AAA50168 | |
| 100 μg | |
| Primary | |
| Detects mouse Plasminogen Kringle 5 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human Angiostatin (Kringle 1-3) is observed. This antibody is expected to show cross-reactivity with mouse plasminogen. | |
| Mouse | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction