missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mouse Plasminogen Kringle 5 Antibody, R&D Systems™

Goat Polyclonal Antibody
€ 191.00 - € 557.00
Specifications
| Antigen | Plasminogen Kringle 5 |
|---|---|
| Concentration | LYOPH |
| Dilution | Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL |
| Applications | Western Blot, Immunohistochemistry |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
18770208
![]() |
R&D Systems
AF742-SP |
25 μg |
€ 191.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18079881
|
R&D Systems
AF742 |
100 μg |
€ 557.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Plasminogen Kringle 5 Polyclonal specifically detects Plasminogen Kringle 5 in Mouse samples. It is validated for Western Blot, Immunohistochemistry.Specifications
| Plasminogen Kringle 5 | |
| Western Blot 0.1 ug/mL, Immunohistochemistry 5-15 ug/mL | |
| Polyclonal | |
| Goat | |
| Mouse | |
| 5340 | |
| E. coli-derived recombinant mouse Plasminogen Kringle 5 Cys481-Ala563 Accession # AAA50168 | |
| Primary | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Detects mouse Plasminogen Kringle 5 in direct ELISAs and Western blots. In direct ELISAs, less than 10% cross-reactivity with recombinant human Angiostatin (Kringle 1-3) is observed. This antibody is expected to show cross-reactivity with mouse plasminogen. |
| LYOPH | |
| Western Blot, Immunohistochemistry | |
| Unconjugated | |
| RUO | |
| AAA50168 | |
| Plg | |
| IgG | |
| Affinity Purified | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title