Learn More
MP Biomedicals™ Bovine Albumin, from New Zealand-sourced Bovine Plasma
Brand: MP Biomedicals™ 0218062010
Additional Details : CAS : 9048-46-8 Weight : 0.01000kg
Description
Certain conformational and primary-sequence epitopes of BSA are suspected allergens in human beef and milk allergies
- From New Zealand-sourced Bovine Plasma
- Microbiological Grade
- ≥97% Total Protein
- Yellow to Brown Powder
- Synonyms: BSA, Bovine albumin, Albumin from bovine serum
- CAS Number: 9048-46-8
- EC Number: 232-936-2
- MDL Number: MFCD00130384
Specifications
≥97% Total Protein | |
10g | |
Powder |
Bovine Albumin | |
Bovine | |
Microbiological Grade |