missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mrc2 Antibody (SR1302), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-22115-50ul
This item is not returnable.
View return policy
Description
Mrc2 Monoclonal antibody specifically detects Mrc2 in Human samples. It is validated for Western Blot
Specifications
| Mrc2 | |
| Monoclonal | |
| Unconjugated | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| SR1302 | |
| Western Blot 1:500-1:2000 | |
| CD280 antigen, CD280C-type mannose receptor 2, CLEC13EUPAR-associated protein, ENDO180C-type lectin domain family 13 member E, Endocytic receptor 180, KIAA0709FLJ35911, Macrophage mannose receptor 2, mannose receptor, C type 2, UPARAPendocytic receptor (macrophage mannose receptor family), urokinase plasminogen activator receptor-associated protein, Urokinase receptor-associated protein, Urokinase-type plasminogen activator receptor-associated protein | |
| A synthesized peptide derived from human Mrc2 (Uniprot #: Q9UBG0) | |
| 50 μg | |
| CD Markers, Cell Biology, Immunology | |
| 9902 | |
| Store at -20° C. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction