missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Mrc2 Antibody (SR1302), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 349.00 - € 449.00
Specifications
| Antigen | Mrc2 |
|---|---|
| Clone | SR1302 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18632625
|
Novus Biologicals
NBP3-22115-100ul |
100 μg |
€ 449.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18609913
|
Novus Biologicals
NBP3-22115-50ul |
50 μg |
€ 349.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Mrc2 Monoclonal antibody specifically detects Mrc2 in Human samples. It is validated for Western BlotSpecifications
| Mrc2 | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human | |
| CD280 antigen, CD280C-type mannose receptor 2, CLEC13EUPAR-associated protein, ENDO180C-type lectin domain family 13 member E, Endocytic receptor 180, KIAA0709FLJ35911, Macrophage mannose receptor 2, mannose receptor, C type 2, UPARAPendocytic receptor (macrophage mannose receptor family), urokinase plasminogen activator receptor-associated protein, Urokinase receptor-associated protein, Urokinase-type plasminogen activator receptor-associated protein | |
| A synthesized peptide derived from human Mrc2 (Uniprot #: Q9UBG0) | |
| Primary | |
| Store at -20° C. Avoid freeze/thaw cycles. |
| SR1302 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| CD Markers, Cell Biology, Immunology | |
| PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
| 9902 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title