missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL37 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-82622-25ul
This item is not returnable.
View return policy
Description
MRPL37 Polyclonal specifically detects MRPL37 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| MRPL37 | |
| Polyclonal | |
| Western Blot 0.4 μg/mL, Immunohistochemistry, Immunohistochemistry-Paraffin 1:20 - 1:50 | |
| 39S ribosomal protein L2, mitochondrial, L2mt, L37mt, MGC878, mitochondrial ribosomal protein L37, MRP-L2MRPL2, MRP-L37, ribosomal protein, mitochondrial, L2, RPML239S ribosomal protein L37, mitochondrial | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Specificity of antibody verified on a Protein Array containing target protein plus 383 other non-specific proteins. | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS (pH 7.2) and 40% Glycerol with 0.02% Sodium Azide | |
| MRPL37 | |
| This antibody was developed against Recombinant Protein corresponding to amino acids:TDGRVFHFLVFQLNTTDLDSNEGVKNLAWVDSDQLLYQHFWCLPVIKKRVVVEPVGPVGFKPETFRKFLALYLHGA | |
| 25 μL | |
| Core ESC Like Genes, Stem Cell Markers | |
| 51253 | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction