missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL48 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-93432-0.1ml
This item is not returnable.
View return policy
Description
MRPL48 Polyclonal antibody specifically detects MRPL48 in Human, Mouse, Rat samples. It is validated for Western Blot
Specifications
| MRPL48 | |
| Polyclonal | |
| Western Blot 1:500-1:2000 | |
| 39S ribosomal protein L48, mitochondrial, CGI-118, FLJ17047, FLJ99260, HSPC290, L48MT, MGC13323, mitochondrial ribosomal protein L48, MRP-L48 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-212 of human MRPL48 (NP_057139.1). MSGTLEKVLCLRNNTIFKQAFSLLRFRTSGEKPIYSVGGILLSISRPYKTKPTHGIGKYKHLIKAEEPKKKKGKVEVRAINLGTDYEYGVLNIHLTAYDMTLAESYAQYVHNLCNSLSIKVEESYAMPTKTIEVLQLQDQGSKMLLDSVLTTHERVVQISGLSATFAEIFLEIIQSSLPEGVRLSVKEHTEEDFKGRFKARPELEELLAKLK | |
| 0.1 mL | |
| Cancer, Endocrinology, Signal Transduction | |
| 51642 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction