missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MRPL48 Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 226.00 - € 470.00
Specifications
| Antigen | MRPL48 |
|---|---|
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18638521
|
Novus Biologicals
NBP2-93432-0.02ml |
0.02 mL |
€ 226.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18654651
|
Novus Biologicals
NBP2-93432-0.1ml |
0.1 mL |
€ 470.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MRPL48 Polyclonal antibody specifically detects MRPL48 in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| MRPL48 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Endocrinology, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 51642 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:2000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat | |
| 39S ribosomal protein L48, mitochondrial, CGI-118, FLJ17047, FLJ99260, HSPC290, L48MT, MGC13323, mitochondrial ribosomal protein L48, MRP-L48 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-212 of human MRPL48 (NP_057139.1). MSGTLEKVLCLRNNTIFKQAFSLLRFRTSGEKPIYSVGGILLSISRPYKTKPTHGIGKYKHLIKAEEPKKKKGKVEVRAINLGTDYEYGVLNIHLTAYDMTLAESYAQYVHNLCNSLSIKVEESYAMPTKTIEVLQLQDQGSKMLLDSVLTTHERVVQISGLSATFAEIFLEIIQSSLPEGVRLSVKEHTEEDFKGRFKARPELEELLAKLK | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title