Learn More
Novus Biologicals™ MS4A14 Antibody (SR4233), Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals™ NBP3-27828-50ul
This item is not returnable.
View return policy
Description
MS4A14 Monoclonal antibody specifically detects MS4A14 in Human samples. It is validated for Western Blot
Specifications
| MS4A14 | |
| Monoclonal | |
| Unconjugated | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 84689 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
| Western Blot | |
| SR4233 | |
| Western Blot 1:500-1:2000 | |
| DKFZp434H092, FLJ32856, membrane-spanning 4-domains subfamily A member 14, membrane-spanning 4-domains, subfamily A, member 14, membrane-spanning 4-domains, subfamily A, member 16, MGC104289, MGC49828, MS4A13, MS4A13 protein, MS4A16, NYD-SP21, testes development-related NYD-SP21, Testis development protein NYD-SP21 | |
| A synthesized peptide derived from human MS4A14 (Uniprot #: Q96JA4) | |
| 50 μL | |
| Primary | |
| Human | |
| Purified |