Learn More
MS4A2 Rabbit, anti-Human, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand:  Proteintech 26808-1-AP-20UL
 View more versions of this product
Product Code. 18025115
  • € 145.00 / 20µL
Description
The MS4A2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MS4A2. This antibody recognizes human, mouse antigen. The MS4A2 antibody has been validated for the following applications: WB, ELISA analysis.
Specifications
Specifications
MS4A2
550 μg/mL
Rabbit
Primary
Human, Mouse
150 kDa
IgG
Western Blot, ELISA
APY, ATOPY, FCER1B, FCERI, IgE Fc receptor subunit beta, IGEL, IGER, IGHER, MS4A1, MS4A2
20 μL
2206
Store at -20°C. Stable for one year after shipment.