missing translation for 'onlineSavingsMsg'
Learn More

MS4A2 Rabbit, anti-Human, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

€ 145.00 - € 410.00

Specifications

Antigen MS4A2
Concentration 550 μg/mL
Applications Western Blot, ELISA
Form Liquid
Host Species Rabbit
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18025115
View Documents
Proteintech
26808-1-AP-20UL
20 μL
€ 145.00
20µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18006695
View Documents
Proteintech
26808-1-AP-150UL
150 μL
€ 410.00
150µL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

The MS4A2 antibody from Proteintech is a rabbit polyclonal antibody generated with recombinant protein of human MS4A2. This antibody recognizes human, mouse antigen. The MS4A2 antibody has been validated for the following applications: WB, ELISA analysis.

Specifications

Specifications

MS4A2
Western Blot, ELISA
Rabbit
2206
IgG
Store at -20°C. Stable for one year after shipment.
550 μg/mL
Liquid
Human, Mouse
APY, ATOPY, FCER1B, FCERI, IgE Fc receptor subunit beta, IGEL, IGER, IGHER, MS4A1, MS4A2
Primary
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.