missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MS4A4A Rabbit anti-Human, Clone: 3F2, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP3-12045-0.025MG
This item is not returnable.
View return policy
Description
MS4A4A Monoclonal antibody specifically detects MS4A4A in Human samples. It is validated for Flow Cytometry, Immunofluorescence, Immunoprecipitation
Specifications
| MS4A4A | |
| Monoclonal | |
| 1 mg/ml | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| 4SPAN1, CD20 antigen-like 1, CD20L1CD20-L1, Fc epsilon receptor beta subunit homolog, Four-span transmembrane protein 1, membrane-spanning 4-domains subfamily A member 4A, membrane-spanning 4-domains, subfamily A, member 4, membrane-spanning 4-domains, subfamily A, member 4A, MS4A4MGC22311, MS4A7 | |
| This antibody was generated by immunizing C57BL/6 mice against mouse cell line 300-19 expressing transfected human membrane-spanning 4-domains subfamily A member 4A (MS4A4A)-GFP. | |
| 0.025 mg | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry, Immunofluorescence, Immunoprecipitation | |
| 3F2 | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 51338 | |
| Store at 4C for up to 3 months. For longer storage, aliquot and store at -20C. | |
| IgG κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction