missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MS4A4A Rabbit anti-Human, Clone: 3F2, Novus Biologicals™
Rabbit Monoclonal Antibody
€ 274.00
Specifications
| Antigen | MS4A4A |
|---|---|
| Clone | 3F2 |
| Dilution | Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation |
| Classification | Monoclonal |
| Conjugate | Unconjugated |
Description
MS4A4A Monoclonal antibody specifically detects MS4A4A in Human samples. It is validated for Flow Cytometry, Immunofluorescence, ImmunoprecipitationSpecifications
| MS4A4A | |
| Flow Cytometry, Immunocytochemistry/Immunofluorescence, Immunoprecipitation | |
| Unconjugated | |
| RUO | |
| PBS | |
| 51338 | |
| IgG κ | |
| Protein A purified |
| 3F2 | |
| Monoclonal | |
| Rabbit | |
| Human | |
| 4SPAN1, CD20 antigen-like 1, CD20L1CD20-L1, Fc epsilon receptor beta subunit homolog, Four-span transmembrane protein 1, membrane-spanning 4-domains subfamily A member 4A, membrane-spanning 4-domains, subfamily A, member 4, membrane-spanning 4-domains, subfamily A, member 4A, MS4A4MGC22311, MS4A7 | |
| This antibody was generated by immunizing C57BL/6 mice against mouse cell line 300-19 expressing transfected human membrane-spanning 4-domains subfamily A member 4A (MS4A4A)-GFP. | |
| Primary |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title