Learn More
Invitrogen™ MSRB3 Recombinant Rabbit Monoclonal Antibody (24GB3550)
Rabbit Recombinant Monoclonal Antibody
Brand:  Invitrogen™ MA557076
 View more versions of this product
Product Code. 30276270
  • € 151.00 / 20µL
Description
MSRB3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
The protein encoded by this gene catalyzes the reduction of methionine sulfoxide to methionine. This enzyme acts as a monomer and requires zinc as a cofactor. Several transcript variants encoding two different isoforms have been found for this gene. One of the isoforms localizes to mitochondria while the other localizes to endoplasmic reticula.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
MSRB3
Recombinant Monoclonal
Unconjugated
MSRB3
D430026P16Rik; DFNB74; I-8-23; methionine sulfoxide reductase B3; methionine-R-sulfoxide reductase B3; Methionine-R-sulfoxide reductase B3, mitochondrial; MsrB2; MsrB3; UNQ1965/PRO4487
Rabbit
Affinity chromatography
RUO
253827, 320183, 680036
-20°C
Liquid
Western Blot, Immunocytochemistry
24GB3550
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Q8BU85, Q8IXL7
MSRB3
A synthesized peptide derived from human MSRB3 (aa 50-192).
20 μL
Primary
Human, Mouse, Rat