missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ MSRB3 Recombinant Rabbit Monoclonal Antibody (24GB3550)
Rabbit Recombinant Monoclonal Antibody
€ 151.00 - € 514.00
Specifications
| Antigen | MSRB3 |
|---|---|
| Clone | 24GB3550 |
| Content And Storage | -20°C |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Recombinant Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30276277
|
Invitrogen™
MA557075 |
100 μL |
€ 514.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30276270
|
Invitrogen™
MA557076 |
20 μL |
€ 151.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MSRB3 Recombinant Monoclonal Antibody for Western Blot, ICC/IF
The protein encoded by this gene catalyzes the reduction of methionine sulfoxide to methionine. This enzyme acts as a monomer and requires zinc as a cofactor. Several transcript variants encoding two different isoforms have been found for this gene. One of the isoforms localizes to mitochondria while the other localizes to endoplasmic reticula.Specifications
| MSRB3 | |
| -20°C | |
| Recombinant Monoclonal | |
| Liquid | |
| IgG | |
| Human, Mouse, Rat | |
| MSRB3 | |
| D430026P16Rik; DFNB74; I-8-23; methionine sulfoxide reductase B3; methionine-R-sulfoxide reductase B3; Methionine-R-sulfoxide reductase B3, mitochondrial; MsrB2; MsrB3; UNQ1965/PRO4487 | |
| MSRB3 | |
| Primary | |
| Affinity chromatography |
| 24GB3550 | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q8BU85, Q8IXL7 | |
| 253827, 320183, 680036 | |
| A synthesized peptide derived from human MSRB3 (aa 50-192). | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title