Learn More
MT-ND1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-94324-0.02ml
This item is not returnable.
View return policy
Description
MT-ND1 Polyclonal antibody specifically detects MT-ND1 in Human, Mouse samples. It is validated for Western Blot
Specifications
| MT-ND1 | |
| Polyclonal | |
| Western Blot 1:500-1:1000 | |
| EC 1.6.5.3, mitochondrially encoded NADH dehydrogenase 1, MTND1, NAD1, NADH dehydrogenase subunit 1, NADH dehydrogenase, subunit 1 (complex I), NADH1, ND1NADH dehydrogenase 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of mouse MT-ND1 (NP_904328.1). MFFINILTLLVPILIAMAFLTLVERKILGYMQLRKGPNIVGPYGILQPFADAMKLFMKEPMRPLTTSMSLFIIAPTLSLTLALSLWVPLPMPHPLINLNL | |
| 0.02 mL | |
| Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| 4535 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
| Western Blot | |
| Unconjugated | |
| PBS (pH 7.3), 50% glycerol | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |