missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MT-ND1 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 218.00 - € 493.00
Specifications
| Antigen | MT-ND1 |
|---|---|
| Dilution | Western Blot 1:500-1:1000 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18641622
|
Novus Biologicals
NBP2-94324-0.02ml |
0.02 mL |
€ 218.00
0.02mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18653162
|
Novus Biologicals
NBP2-94324-0.1ml |
0.1 mL |
€ 493.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
MT-ND1 Polyclonal antibody specifically detects MT-ND1 in Human, Mouse samples. It is validated for Western BlotSpecifications
| MT-ND1 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer, Endocrinology, Neurodegeneration, Neuroscience, Signal Transduction | |
| PBS (pH 7.3), 50% glycerol | |
| 4535 | |
| IgG | |
| Affinity purified |
| Western Blot 1:500-1:1000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| EC 1.6.5.3, mitochondrially encoded NADH dehydrogenase 1, MTND1, NAD1, NADH dehydrogenase subunit 1, NADH dehydrogenase, subunit 1 (complex I), NADH1, ND1NADH dehydrogenase 1 | |
| A synthetic peptide corresponding to a sequence within amino acids 1-100 of mouse MT-ND1 (NP_904328.1). MFFINILTLLVPILIAMAFLTLVERKILGYMQLRKGPNIVGPYGILQPFADAMKLFMKEPMRPLTTSMSLFIIAPTLSLTLALSLWVPLPMPHPLINLNL | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title