missing translation for 'onlineSavingsMsg'
Learn More

MUC13 Antibody - BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

Brand:  Novus Biologicals NBP2-24566SS

 View more versions of this product

Product Code. 18136165

  • € 226.00 / 25µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

MUC13 Polyclonal specifically detects MUC13 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

MUC13
Polyclonal
Unconjugated
PBS with 0.05% Sodium Azide
MUC13
A portion of amino acids 480-512 of human Mucin 13 was used as the immunogen.
0.025 mg
Primary
The amino acid sequence used as immunogen is 100% homologous in human.
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.
Western Blot, Immunohistochemistry (Paraffin)
1.0 mg/mL
Western Blot 0.25-0.50 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml
Down-regulated in colon cancer 1FLJ20063, DRCC1, MUC-13, mucin 13, cell surface associated, mucin 13, epithelial transmembrane, mucin-13, RECC
Rabbit
Affinity Purified
RUO
56667
Human
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.

For Research Use Only