missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC13 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 226.00 - € 338.00
Specifications
| Antigen | MUC13 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Western Blot 0.25-0.50 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml |
| Applications | Western Blot, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18136165
|
Novus Biologicals
NBP2-24566SS |
0.025 mg |
€ 226.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18781384
|
Novus Biologicals
NBP2-24566 |
€ 338.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||||
Description
MUC13 Polyclonal specifically detects MUC13 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
| MUC13 | |
| Western Blot 0.25-0.50 ug/ml, Immunohistochemistry-Paraffin 5 ug/ml | |
| Polyclonal | |
| Rabbit | |
| Human | |
| Down-regulated in colon cancer 1FLJ20063, DRCC1, MUC-13, mucin 13, cell surface associated, mucin 13, epithelial transmembrane, mucin-13, RECC | |
| MUC13 | |
| IgG | |
| Affinity Purified | |
| The amino acid sequence used as immunogen is 100% homologous in human. |
| 1.0 mg/mL | |
| Western Blot, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| RUO | |
| PBS with 0.05% Sodium Azide | |
| 56667 | |
| A portion of amino acids 480-512 of human Mucin 13 was used as the immunogen. | |
| Primary | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title