Learn More
MUC13 Antibody - BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP2-24566
 View more versions of this product
Product Code. 18781384
  • € 338.00 / 0.10mg
Description
MUC13 Polyclonal specifically detects MUC13 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Polyclonal
Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles.