missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-23576SS
This item is not returnable.
View return policy
Description
MUC2 Polyclonal antibody specifically detects MUC2 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Flow (Intracellular)
Specifications
| MUC2 | |
| Polyclonal | |
| Unconjugated | |
| PBS | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry (Intracellular Staining) | |
| 1.0 mg/mL | |
| Flow Cytometry 1.0 mg/mL, Immunohistochemistry 1:200, Immunocytochemistry/ Immunofluorescence 1 μg/mL, Immunohistochemistry-Paraffin 1:200, Flow (Intracellular) 1.0 mg/mL | |
| Intestinal mucin-2, MLP, MUC2, MUC-2, mucin 2, intestinal/tracheal, mucin 2, oligomeric mucus/gel-forming, mucin-2, SMUC | |
| Synthetic peptide made to an internal portion of the human MUC2 protein (between amino acids 4900-4950) [UniProt Q02817] | |
| 0.025 mg | |
| Cancer, Cellular Markers, Inflammation, Signal Transduction | |
| 4583 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction