missing translation for 'onlineSavingsMsg'
Learn More
Learn More
MUC2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 208.00 - € 450.00
Specifications
| Antigen | MUC2 |
|---|---|
| Concentration | 1.0 mg/mL |
| Dilution | Flow Cytometry 1.0 mg/mL, Immunohistochemistry 1:200, Immunocytochemistry/ Immunofluorescence 1 μg/mL, Immunohistochemistry-Paraffin 1:200, Flow (Intracellular) 1.0 mg/mL |
| Applications | Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry (Intracellular Staining) |
| Classification | Polyclonal |
| Produktkod | Brand | Quantity | Pris | Kvantitet och tillgänglighet | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktkod | Brand | Quantity | Pris | Kvantitet och tillgänglighet | |||||
|
18650714
|
Novus Biologicals
NBP3-23576 |
0.1 mg |
€ 450.00
0.10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18672684
|
Novus Biologicals
NBP3-23576SS |
0.025 mg |
€ 208.00
0.02mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beskrivning
MUC2 Polyclonal antibody specifically detects MUC2 in Human, Mouse samples. It is validated for Flow Cytometry, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence, Immunohistochemistry (Paraffin), Flow (Intracellular)Specifikationer
| MUC2 | |
| Flow Cytometry 1.0 mg/mL, Immunohistochemistry 1:200, Immunocytochemistry/ Immunofluorescence 1 μg/mL, Immunohistochemistry-Paraffin 1:200, Flow (Intracellular) 1.0 mg/mL | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Mouse | |
| Intestinal mucin-2, MLP, MUC2, MUC-2, mucin 2, intestinal/tracheal, mucin 2, oligomeric mucus/gel-forming, mucin-2, SMUC | |
| Synthetic peptide made to an internal portion of the human MUC2 protein (between amino acids 4900-4950) [UniProt Q02817] | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
| 1.0 mg/mL | |
| Flow Cytometry, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Flow Cytometry (Intracellular Staining) | |
| Unconjugated | |
| Rabbit | |
| Cancer, Cellular Markers, Inflammation, Signal Transduction | |
| PBS | |
| 4583 | |
| IgG | |
| Immunogen affinity purified |
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel