Learn More
Shamrock™ Multi-Purpose Blank Tyvek Labels
Provides extra strength, adhesion and permanent marking for your laboratory needs. Shamrock™ Multi-Purpose Blank Tyvek Labels won't discolor, crack or tear, adhere to almost any surface and are available in a range of sizes.
Brand: Shamrock™ STV-4
Additional Details : Weight : 0.80000kg
Description
- Tear-proof material, long-lasting adhesive
- Does not discolor over time
- Permanent marking with ballpoint pen, paper pencil and felt marker
- Labels pre-cut on roll
Specifications
Permanent | |
Tyvek | |
For Unique and demanding applications | |
38 x 76 mm |
White | |
Multi-Purpose Blank Tyvek Label | |
38 x 76 mm |