missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Muscleblind-like 1 Antibody (1H1), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-50062
This item is not returnable.
View return policy
Description
Muscleblind-like 1 Monoclonal antibody specifically detects Muscleblind-like 1 in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
Specifications
| Muscleblind-like 1 | |
| Monoclonal | |
| 1 mg/mL | |
| Western Blot 1:1000 - 1:2000, Immunohistochemistry 1:1000, Immunocytochemistry/ Immunofluorescence 1:1000 | |
| Q9NR56 | |
| Mouse | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse (Negative), Rat (Negative) | |
| Purified |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| 1H1 | |
| Unconjugated | |
| 50% PBS, 50% glycerol | |
| DKFZp686P06174, EXP40, EXP42, EXPKIAA0428EXP35, MBNL, muscleblind (Drosophila)-like, muscleblind-like (Drosophila), muscleblind-like protein 1, Triplet-expansion RNA-binding protein | |
| Full-length recombinant human muscleblind-like 1 protein expressed in and purified from E. coli. [UniProt# Q9NR56] | |
| 0.1 mL | |
| Chromatin Research, DNA replication Transcription Translation and Splicing | |
| 4154 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction