missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Muscleblind-like 1 Antibody (1H1), Novus Biologicals™
Mouse Monoclonal Antibody
€ 220.00 - € 464.00
Specifications
| Antigen | Muscleblind-like 1 |
|---|---|
| Clone | 1H1 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:1000 - 1:2000, Immunohistochemistry 1:1000, Immunocytochemistry/ Immunofluorescence 1:1000 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18674626
|
Novus Biologicals
NBP2-50062SS |
0.025 mL |
€ 220.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18614767
|
Novus Biologicals
NBP2-50062 |
0.1 mL |
€ 464.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Muscleblind-like 1 Monoclonal antibody specifically detects Muscleblind-like 1 in Human, Mouse (Negative), Rat (Negative) samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| Muscleblind-like 1 | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Chromatin Research, DNA replication Transcription Translation and Splicing | |
| 50% PBS, 50% glycerol | |
| DKFZp686P06174, EXP40, EXP42, EXPKIAA0428EXP35, MBNL, muscleblind (Drosophila)-like, muscleblind-like (Drosophila), muscleblind-like protein 1, Triplet-expansion RNA-binding protein | |
| Full-length recombinant human muscleblind-like 1 protein expressed in and purified from E. coli. [UniProt# Q9NR56] | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 1H1 | |
| Western Blot 1:1000 - 1:2000, Immunohistochemistry 1:1000, Immunocytochemistry/ Immunofluorescence 1:1000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse (Negative), Rat (Negative) | |
| Q9NR56 | |
| 4154 | |
| IgG1 | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title